2-Bromo-4-(trifluoromethyl)benzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Bromo-4-(trifluoromethyl)benzoic acid, 2-Bromo-4-(trifluoromethl)benzoic acid

B5334-1G B5334-5G
TCB5334-1GEA 80.58 CAD
TCB5334-1G TCB5334-5G
2-Bromo-4-(trifluoromethyl)benzoic acid ≥98.0% (by GC, titration analysis)
2-Bromo-4-(trifluoromethyl)benzoic acid
Formula: C₈H₄BrF₃O₂
MW: 269.02
MDL Number: MFCD07368837
CAS Number: 328-89-2
EINECS: 206-339-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR