ω,3-Dibromoacetophenone ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

B4125-25G B4125-5G
TCB4125-25GEA 598.82 CAD
TCB4125-25G TCB4125-5G
ω,3-Dibromoacetophenone ≥98.0% (by titrimetric analysis)
ω,3-Dibromoacetophenone
Formula: C₈H₆Br₂O
MDL Number: MFCD00082698
CAS Number: 18523-22-3
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 49.0 to 53.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR