3-Ethynylanisole ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 1-Ethynyl-3-methoxybenzene, (3-Methoxyphenyl)ethyne, (3-Methoxyphenyl)acetylene, 1-Methoxy-3-ethynylbenzene

E0627-1G E0627-5G
TCE0627-1GEA 83.74 CAD
TCE0627-1G TCE0627-5G
3-Ethynylanisole ≥97.0% (by GC)
3-Ethynylanisole
Formula: C₉H₈O
MW: 132.16
Boiling Pt: 90°C
Flash Pt: 77°C
Storage Temperature: Refrigerator
MDL Number: MFCD00160810
CAS Number: 768-70-7

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR