1-Bromo-2-ethynylbenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2'-Bromophenylacetylene, 2-Bromophenylacetylene

B4608-1G B4608-5G
TCB4608-1GEA 195.92 CAD
TCB4608-1G TCB4608-5G
1-Bromo-2-ethynylbenzene ≥98.0% (by GC)
1-Bromo-2-ethynylbenzene
Formula: C₈H₅Br
MW: 181.03
Boiling Pt: 214
Melting Pt: 52
Density: 1.4434
Flash Pt: 63°C
MDL Number: MFCD01863358
CAS Number: 766-46-1

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR