4-(Trifluoromethoxy)phenyl isothiocyanate ≥98.0% (by GC)

Supplier: TCI America
Danger

T3341-1G T3341-5G
TCT3341-1GEA 56.25 CAD
TCT3341-1G TCT3341-5G
4-(Trifluoromethoxy)phenyl isothiocyanate ≥98.0% (by GC)
4-(Trifluoromethoxy)phenyl isothiocyanate
Formula: C₈H₄F₃NOS
Boiling Pt: 88°C
Flash Pt: 107°C
Storage Temperature: Refrigerator
MDL Number: MFCD00042412
CAS Number: 64285-95-6
UN: 1760
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR