4-Amino-2-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
A3164-25G
A3164-5G
TCA3164-25GEA
578.28
CAD
TCA3164-25G
TCA3164-5G
4-Amino-2-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
4-Amino-2-fluorobenzoic acid
Formula:
C₇H₆FNO₂ MW: 155.13 |
MDL Number:
MFCD01569397 CAS Number: 446-31-1 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...