3-Chloro-2-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
C3261-1G
C3261-5G
TCC3261-1GEA
178.54
CAD
TCC3261-1G
TCC3261-5G
3-Chloro-2-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
3-Chloro-2-methoxybenzoic acid
Formula:
C₈H₇ClO₃ MW: 186.59 Storage Temperature: Refrigerator |
MDL Number:
MFCD00179306 CAS Number: 3260-93-3 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 120.0 to 124.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...