2,4-Dichlorobenzhydrazide ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D4867-1G D4867-5G
TCD4867-1GEA 48.98 CAD
TCD4867-1G TCD4867-5G
2,4-Dichlorobenzhydrazide ≥98.0% (by GC, titration analysis)
2,4-Dichlorobenzhydrazide
Formula: C₇H₆Cl₂N₂O
MDL Number: MFCD00025113
CAS Number: 5814-06-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 167.0 to 171.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR