Magnesium stearate

Supplier: TCI America
Warning

S0238-250G S0238-25G
TCS0238-250GEA 55.3 CAD
TCS0238-250G TCS0238-25G
Magnesium stearate
Magnesium stearate
Formula: C₃₆H₇₀MgO₄
MDL Number: MFCD00036391
CAS Number: 557-04-0
EINECS: 209-150-3
Merck Index: 14,05690

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Content of Magnesium 4.00 to 5.00 %
Palmitic acid (composition of fatty acid) max. 40.0 %
Stearic acid (composition of fatty acid) min. 60.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR