4-Methyl-2-phenylpyridine ≥97.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
M2952-1G
M2952-200MG
TCM2952-1GEA
420.28
CAD
TCM2952-1G
TCM2952-200MG
4-Methyl-2-phenylpyridine ≥97.0% (by GC, titration analysis)
4-Methyl-2-phenylpyridine
Formula:
C₁₂H₁₁N Boiling Pt: 115°C |
MDL Number:
MFCD00087695 CAS Number: 3475-21-6 EINECS: 222-448-8 |
Specification Test Results
Appearance | White to Light yellow powder to lump |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 47.0 to 51.0 deg-C |
melting range | max. 5.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...