4-(Trifluoromethyl)benzamide ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
T3474-25G
T3474-5G
TCT3474-25GEA
240.16
CAD
TCT3474-25G
TCT3474-5G
4-(Trifluoromethyl)benzamide ≥98.0% (by GC)
4-(Trifluoromethyl)benzamide
Formula:
C₈H₆F₃NO MW: 189.14 |
MDL Number:
MFCD00007998 CAS Number: 1891-90-3 EINECS: 217-571-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 183.0 to 187.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...