3-Fluoro-5-hydroxypyridine ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
F1020-1G
F1020-200MG
TCF1020-1GEA
206.66
CAD
TCF1020-1G
TCF1020-200MG
3-Fluoro-5-hydroxypyridine ≥98.0% (by GC)
3-Fluoro-5-hydroxypyridine
Formula:
C₅H₄FNO Storage Temperature: Refrigerator |
MDL Number:
MFCD03095199 CAS Number: 209328-55-2 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 162.0 to 166.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...