2-Chloro-6-fluoropyridine ≥98.0% (by GC)

Supplier: TCI America
Danger

C3186-25G C3186-5G
TCC3186-25GEA 188.02 CAD
TCC3186-25G TCC3186-5G
2-Chloro-6-fluoropyridine ≥98.0% (by GC)
2-Chloro-6-fluoropyridine
Formula: C₅H₃ClFN
Storage Temperature: Refrigerator
MDL Number: MFCD00077483
CAS Number: 20885-12-5

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Melting point 31.0 to 35.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR