1-Bromo-4-fluoro-2-iodobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

B4506-25G B4506-5G
76219-398EA 306.31 CAD
76219-398 76219-400
1-Bromo-4-fluoro-2-iodobenzene ≥98.0% (by GC)
1-Bromo-4-fluoro-2-iodobenzene
Formula: C₆H₃BrFI
MDL Number: MFCD00042578
CAS Number: 202865-72-3

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR