5-Bromo-2,3-difluorophenol ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-Bromo-3,4-difluoro-5-hydroxybenzene

B4490-25G B4490-5G
TCB4490-25GEA 265.44 CAD
TCB4490-25G TCB4490-5G
5-Bromo-2,3-difluorophenol ≥98.0% (by GC)
5-Bromo-2,3-difluorophenol
Formula: C₆H₃BrF₂O
MW: 208.99 g/mol
MDL Number: MFCD01631350
CAS Number: 186590-26-1

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR