2-Bromo-6-fluoroaniline ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B4803-25G
B4803-5G
TCB4803-25GEA
346.02
CAD
TCB4803-25G
TCB4803-5G
2-Bromo-6-fluoroaniline ≥98.0% (by GC)
2-Bromo-6-fluoroaniline
Formula:
C₆H₅BrFN MW: 190.01 |
MDL Number:
MFCD01310982 CAS Number: 65896-11-9 EINECS: 464-930-2 |
Specification Test Results
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...