α-Chloro-2,5-difluorotoulene ≥98.0% (by GC)
Supplier: TCI America
|
Danger
|
D4726-25G
D4726-5G
TCD4726-25GEA
638
CAD
TCD4726-25G
TCD4726-5G
α-Chloro-2,5-difluorotoulene ≥98.0% (by GC)
α-Chloro-2,5-difluorotoulene
Formula:
C₇H₅ClF₂ Boiling Pt: 74°C |
MDL Number:
MFCD01090989 CAS Number: 495-07-8 UN: 3265 ADR: 8,III |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...