2,3,5,6-Tetrabromo-p-xylene ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1,2,4,5-Tetrabromo-3,6-dimethylbenzene

T3202-1G T3202-5G
TCT3202-1GEA 137.14 CAD
TCT3202-1G TCT3202-5G
2,3,5,6-Tetrabromo-p-xylene ≥98.0% (by GC)
2,3,5,6-Tetrabromo-p-xylene
Formula: C₈H₆Br₄
MW: 421.75 g/mol
MDL Number: MFCD00010353
CAS Number: 23488-38-2
EINECS: 245-688-5

Order Now

Specification Test Results

Appearance White to Light gray to Light yellow powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR