3',5'-Dichloroacetophenone ≥98.0% (by GC)

Supplier: TCI America
Warning

D5111-1G D5111-5G
TCD5111-1GEA 63.2 CAD
TCD5111-1G TCD5111-5G
3',5'-Dichloroacetophenone ≥98.0% (by GC)
3',5'-Dichloroacetophenone
Formula: C₈H₆Cl₂O
Boiling Pt: 135°C
MDL Number: MFCD00045189
CAS Number: 14401-72-0

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR