3-Methyladenine

Supplier: TCI America
Warning

M2518-1G M2518-200MG
TCM2518-1GEA 778.94 CAD
TCM2518-1G TCM2518-200MG
3-Methyladenine
3-Methyladenine
Formula: C₆H₇N₅
MDL Number: MFCD00010531
CAS Number: 5142-23-4
EINECS: 225-908-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR