3-Cyclopentylpropionic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C2934-25G C2934-5G
TCC2934-25GEA 143.78 CAD
TCC2934-25G TCC2934-5G
3-Cyclopentylpropionic acid ≥98.0% (by GC, titration analysis)
3-Cyclopentylpropionic acid
Formula: C₈H₁₄O₂
Boiling Pt: 123°C
Flash Pt: 47°C
MDL Number: MFCD00001392
CAS Number: 140-77-2
EINECS: 205-433-0
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR