2',6'-Dimethoxyacetophenone ≥98.0% (by GC)

Supplier: TCI America

D4849-1G D4849-5G
76219-720EA 33.18 CAD
76219-720 76219-722
2',6'-Dimethoxyacetophenone ≥98.0% (by GC)
2',6'-Dimethoxyacetophenone
Formula: C₁₀H₁₂O₃
Boiling Pt: 136°C
MDL Number: MFCD00008729
CAS Number: 2040-04-2
EINECS: 218-034-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 68.0 to 72.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR