4'-Chlorochalcone 98,0 GC_ASSAY_METHOD
Supplier: TCI America
|
Warning
|
Synonyms:
1-(4-Chlorophenyl)-3-phenylprop-2-en-1-one, 4'-Chlorobenzylidenacetophenone
C3259-25G
C3259-5G
TCC3259-25GEA
317.58
CAD
TCC3259-25G
TCC3259-5G
4'-Chlorochalcone 98,0 GC_ASSAY_METHOD
4'-Chlorochalcone
Formula:
C₁₅H₁₁ClO MW: 242.7 |
MDL Number:
MFCD00016343 CAS Number: 956-02-5 |
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 95.0 and le 99.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...