3-Bromobenzylamine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Danger
|
B4676-25G
B4676-5G
TCB4676-25GEA
360.4
CAD
TCB4676-25G
TCB4676-5G
3-Bromobenzylamine ≥98.0% (by titrimetric analysis)
3-Bromobenzylamine
Formula:
C₇H₈BrN Boiling Pt: 245°C Storage Temperature: Refrigerator |
MDL Number:
MFCD01026119 CAS Number: 10269-01-9 UN: 2735 ADR: 8,III |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...