Triphenylene ≥98.0% (by GC), purified by sublimation

Supplier: TCI America
Warning

Synonyms: 9,10-Benzophenanthrene, Isochrysene

T3267-1G T3267-200MG
TCT3267-1GEA 776.89 CAD
TCT3267-1G TCT3267-200MG
Triphenylene ≥98.0% (by GC), purified by sublimation
Triphenylene
Formula: C₁₈H₁₂
MW: 228.29 g/mol
Boiling Pt: 438
Melting Pt: 199
MDL Number: MFCD00001108
CAS Number: 217-59-4
EINECS: 205-922-9
Merck Index: 14,09740

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 197.0 to 201.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR