Ammonium thiocyanate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Thiocyanic acid ammonium salt, Ammonium rhodanide, Ammonium sulphocyanate, Ammonium sulfocyanate

A3088-25G A3088-500G
TCA3088-25GEA 42.66 CAD
TCA3088-25G TCA3088-500G
Ammonium thiocyanate ≥98.0% (by titrimetric analysis)
Ammonium thiocyanate
Formula: NH₄SCN
Melting Pt: 150°C
MDL Number: MFCD00011428
CAS Number: 1762-95-4
EINECS: 217-175-6
Merck Index: 14,00561

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (NH4SCN method) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR