Potassium disulfite ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Danger

P2480-25G P2480-500G
TCP2480-25GEA 45.82 CAD
TCP2480-25G TCP2480-500G
Potassium disulfite ≥97.0% (by titrimetric analysis)
di-Potassium disulphite
Formula: K₂O₅S₂
Melting Pt: 190°C
MDL Number: MFCD00167605
CAS Number: 16731-55-8
EINECS: 240-795-3
Merck Index: 14,07645

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR