Isoquinoline-1-amine ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
A2817-1G
A2817-5G
TCA2817-1GEA
134.3
CAD
TCA2817-1G
TCA2817-5G
Isoquinoline-1-amine ≥98.0% (by GC)
Isoquinoline-1-amine
Formula:
C₉H₈N₂ Boiling Pt: 166°C |
MDL Number:
MFCD00024137 CAS Number: 1532-84-9 EINECS: 216-243-2 |
Specification Test Results
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 119.0 to 123.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...