2-Fluoro-3-nitropyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

F0982-25G F0982-5G
TCF0982-25GEA 268.6 CAD
TCF0982-25G TCF0982-5G
2-Fluoro-3-nitropyridine ≥98.0% (by GC)
2-Fluoro-3-nitropyridine
Formula: C₅H₃FN₂O₂
Boiling Pt: 110°C
Melting Pt: 18°C
MDL Number: MFCD03095068
CAS Number: 1480-87-1

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR