2-(Difluoromethoxy)benzaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

D4837-1G D4837-5G
76219-716EA 64.78 CAD
76219-716 76219-718
2-(Difluoromethoxy)benzaldehyde ≥98.0% (by GC)
2-(Difluoromethoxy)benzaldehyde
Formula: C₈H₆F₂O₂
Boiling Pt: 83°C
Flash Pt: 107°C
MDL Number: MFCD00042251
CAS Number: 71653-64-0

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR