Benzyl anthranilate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B5249-1G B5249-5G
TCB5249-1GEA 60.04 CAD
TCB5249-1G TCB5249-5G
Benzyl anthranilate ≥98.0% (by GC, titration analysis)
Benzyl anthranilate
Formula: C₁₄H₁₃NO₂
MW: 227.26
MDL Number: MFCD00060605
CAS Number: 82185-41-9

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 76.0 to 80.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR