2-Chloro-4-methoxypyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

C3024-1G C3024-5G
TCC3024-1GEA 60.04 CAD
TCC3024-1G TCC3024-5G
2-Chloro-4-methoxypyridine ≥98.0% (by GC)
2-Chloro-4-methoxypyridine
Formula: C₆H₆ClNO
MW: 143.57
Boiling Pt: 230°C
Density: 1.258
MDL Number: MFCD02093951
CAS Number: 17228-69-2

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR