2,3-Dichlorophenyl isothiocyanate ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 2,3-Dichlorophenylisothiocyanate, 1,2-Dichloro-3-isothiocyanatobenzene

D4923-25G D4923-5G
TCD4923-25GEA 293.88 CAD
TCD4923-25G TCD4923-5G
2,3-Dichlorophenyl isothiocyanate ≥98.0% (by GC)
2,3-Dichlorophenyl isothiocyanate
Formula: C₇H₃Cl₂NS
Boiling Pt: 258°C
MDL Number: MFCD00041048
CAS Number: 6590-97-2
UN: 1760
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR