2-Amino-5-chloro-3-iodopyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A3069-1G A3069-5G
TCA3069-1GEA 123.24 CAD
TCA3069-1G TCA3069-5G
2-Amino-5-chloro-3-iodopyridine ≥98.0% (by GC, titration analysis)
2-Amino-5-chloro-3-iodopyridine
Formula: C₅H₄ClIN₂
MDL Number: MFCD07644578
CAS Number: 211308-81-5

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 96.0 to 100.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR