2-Chloro-4-iodobenzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

C3281-1G C3281-5G
TCC3281-1GEA 94.8 CAD
TCC3281-1G TCC3281-5G
2-Chloro-4-iodobenzoic acid ≥98.0% (by titrimetric analysis)
2-Chloro-4-iodobenzoic acid
Formula: C₇H₄ClIO₂
MDL Number: MFCD05863523
CAS Number: 145343-76-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 165.0 to 169.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR