Titanium(IV) chloride ∼19% ∼1.0 mol/L in toluene

Supplier: TCI America
Danger

T3238-100ML T3238-500ML
TCT3238-100MLEA 53.72 CAD
TCT3238-100ML TCT3238-500ML
Titanium(IV) chloride ∼19% ∼1.0 mol/L in toluene
Titanium (IV) chloride
Formula: Cl₄Ti
Flash Pt: 8°C
Storage Temperature: Refrigerator
MDL Number: MFCD00011267
CAS Number: 7550-45-0
EINECS: 231-441-9
UN: 3286
ADR: 3,II
Merck Index: 14,09478

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Concentration (Argentmetric titration) 19.0 to 22.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR