2,4-Dibromopyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

D3862-25G D3862-5G
TCD3862-25GEA 431.34 CAD
TCD3862-25G TCD3862-5G
2,4-Dibromopyridine ≥98.0% (by GC)
2,4-Dibromopyridine
Formula: C₅H₃Br₂N
Boiling Pt: 238°C
MDL Number: MFCD01859720
CAS Number: 58530-53-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 38.0 to 42.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR