Aluminium bromide, anhydrous ≥95.0% (by titrimetric analysis)
Supplier: TCI America
|
Danger
|
A2692-25G
A2692-5G
TCA2692-25GEA
253.83
CAD
TCA2692-25G
TCA2692-5G
Aluminium bromide, anhydrous ≥95.0% (by titrimetric analysis)
Aluminium bromide
Formula:
AlBr₃ Boiling Pt: 256°C Melting Pt: 98°C |
MDL Number:
MFCD00003421 CAS Number: 7727-15-3 EINECS: 231-779-7 UN: 1725 ADR: 8,II Merck Index: 14,00332 |
Specification Test Results
Appearance | White to Yellow to Orange powder to crystal |
Purity (Chelometric Titration) | min. 95.0 % |
Purity (Argentmetric Titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...