3-Nitro-2,6-lutidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
N0997-25G
N0997-5G
TCN0997-25GEA
526.14
CAD
TCN0997-25G
TCN0997-5G
3-Nitro-2,6-lutidine ≥98.0% (by GC, titration analysis)
3-Nitro-2,6-lutidine
Formula:
C₇H₈N₂O₂ Boiling Pt: 105°C Flash Pt: 37°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00023503 CAS Number: 15513-52-7 EINECS: 239-543-5 UN: 1325 ADR: 4.1,III |
Specification Test Results
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 36.0 to 40.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...