2-Fluoro-5-nitropyridine ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
F0983-25G
F0983-5G
TCF0983-25GEA
164.32
CAD
TCF0983-25G
TCF0983-5G
2-Fluoro-5-nitropyridine ≥98.0% (by GC)
2-Fluoro-5-nitropyridine
Formula:
C₅H₃FN₂O₂ Boiling Pt: 87°C Melting Pt: 18°C |
MDL Number:
MFCD03095059 CAS Number: 456-24-6 EINECS: 207-260-6 |
Specification Test Results
Appearance | White or Colorless to Yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...