5-Chloroquinoline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C2997-1G C2997-5G
TCC2997-1GEA 100.8 CAD
TCC2997-1G TCC2997-5G
5-Chloroquinoline ≥98.0% (by GC, titration analysis)
5-Chloroquinoline
Formula: C₉H₆ClN
Boiling Pt: 263°C
Storage Temperature: Refrigerator
MDL Number: MFCD00239413
CAS Number: 635-27-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR