8-Quinolinecarbaldehyde ≥95.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Quinoline-8-carbaldehyde, 8-Formylquinoline, 8-Quinolinecarboxaldehyde, Quinoline-8-carboxaldehyde

Q0096-1G Q0096-5G
TCQ0096-1GEA 184.86 CAD
TCQ0096-1G TCQ0096-5G
8-Quinolinecarbaldehyde ≥95.0% (by GC)
8-Quinolinecarbaldehyde
Formula: C₁₀H₇NO
MDL Number: MFCD00805837
CAS Number: 38707-70-9

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 92.0 to 97.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR