2,4-Dimethylbenzonitrile ≥98.0% (by GC)

Supplier: TCI America
Warning

D4495-25G D4495-5G
TCD4495-25GEA 319.16 CAD
TCD4495-25G TCD4495-5G
2,4-Dimethylbenzonitrile ≥98.0% (by GC)
2,4-Dimethylbenzonitrile
Formula: C₉H₉N
Boiling Pt: 109°C
MDL Number: MFCD00016377
CAS Number: 21789-36-6
EINECS: 244-580-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR