2-Fluoro-5-iodotoluene ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-Fluoro-4-iodo-2-methylbenzene, 2-Fluoro-5-iodo-1-methylbenzene

F1004-25G F1004-5G
TCF1004-25GEA 129.56 CAD
TCF1004-25G TCF1004-5G
2-Fluoro-5-iodotoluene ≥98.0% (by GC)
2-Fluoro-5-iodotoluene
Formula: C₇H₆FI
MW: 236.03 g/mol
Boiling Pt: 87°C
MDL Number: MFCD00013710
CAS Number: 452-68-6
EINECS: 207-206-1

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR