2,5-Dibromo-α,α,α-trifluorotoluene ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1,4-Dibromo-2-(trifluoromethyl)benzene, 2,5-Dibromobenzotrifluoride

D4769-25G D4769-5G
TCD4769-25GEA 282.82 CAD
TCD4769-25G TCD4769-5G
2,5-Dibromo-α,α,α-trifluorotoluene ≥98.0% (by GC)
2,5-Dibromo-α,α,α-trifluorotoluene
Formula: C₇H₃Br₂F₃
Boiling Pt: 207°C
MDL Number: MFCD00013554
CAS Number: 7657-09-2
EINECS: 231-620-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %
Melting point 48.0 to 52.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR