2,6-Diiodo-4-nitroaniline ≥98.0% (by GC)

Supplier: TCI America
Warning

D4736-25G D4736-5G
TCD4736-25GEA 944.84 CAD
TCD4736-25G TCD4736-5G
2,6-Diiodo-4-nitroaniline ≥98.0% (by GC)
2,6-Diiodo-4-nitroaniline
Formula: C₆H₄I₂N₂O₂
MDL Number: MFCD00007683
CAS Number: 5398-27-6
EINECS: 226-429-5

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 254.0 to 258.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR