2-Iodobenzaldehyde ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
I0610-1G
I0610-5G
TCI0610-1GEA
151.68
CAD
TCI0610-1G
TCI0610-5G
2-Iodobenzaldehyde ≥98.0% (by GC)
2-Iodobenzaldehyde
Formula:
C₇H₅IO Boiling Pt: 102°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00039570 CAS Number: 26260-02-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 37.0 to 41.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...