2-Chloro-6-nitrophenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C3074-1G C3074-5G
TCC3074-1GEA 52.14 CAD
TCC3074-1G TCC3074-5G
2-Chloro-6-nitrophenol ≥98.0% (by GC, titration analysis)
2-Chloro-6-nitrophenol
Formula: C₆H₄ClNO₃
MW: 173.56
MDL Number: MFCD00024233
CAS Number: 603-86-1
EINECS: 210-061-7

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 69.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR