2-Phenoxybenzaldehyde ≥96.0% (by GC)

Supplier: TCI America

P2475-1G P2475-5G
TCP2475-1GEA 94.8 CAD
TCP2475-1G TCP2475-5G
2-Phenoxybenzaldehyde ≥96.0% (by GC)
2-Phenoxybenzaldehyde
Formula: C₁₃H₁₀O₂
Boiling Pt: 153°C
MDL Number: MFCD00800666
CAS Number: 19434-34-5
EINECS: 243-061-0

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR