3-Ethoxybenzaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

E1133-25G E1133-5G
TCE1133-25GEA 237 CAD
TCE1133-25G TCE1133-5G
3-Ethoxybenzaldehyde ≥98.0% (by GC)
3-Ethoxybenzaldehyde
Formula: C₉H₁₀O₂
Boiling Pt: 245°C
Flash Pt: 110°C
MDL Number: MFCD00016606
CAS Number: 22924-15-8
EINECS: 245-333-4

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR